Merck
  • Morphological transitions of Bacillus subtilis in the presence of food-grade lipidic nanoemulsions.
Morphological transitions of Bacillus subtilis in the presence of food-grade lipidic nanoemulsions.
Journal of food science (2020-03-10)
Neeru Singh, Afzal Hussain, Sandeep Kumar Singh
ABSTRACT
The present study aims to study the antibacterial activity of food-grade lipidic nanoemulsion (noncationized/cationized) against Bacillus subtilis (BS). Bactericidal activity was ascertained by studying the morphological transitions on BS using transmission electron microscopy (TEM), atomic force microscopy (AFM), and scanning electron microscopy (SEM). Morphological changes were witnessed by cell wall breakage, oozing out of cellular contents, loss of cell turgidity and contour. Furthermore, aggregation of cationic nanoemulsion (CaNM) was preferentially observed at apical side of BS construing comparatively more electrostatic attraction between electronegative apical side and CaNM. Resistance response of BS exhibited by apical cell-wall thickening was not able to protect the bacteria due to leakage of cellular content. AFM corroborated its importance in bacteriology, wherein the fragmented cell wall can be "piece-by-piece" identified and sutured back to its appropriate vacant places, thereby, completing the cell wall contour of the ghost cell. Such postmortem analysis of bacterial cell using AFM studies can throw light toward mechanism of cell fragmentation of bacterial cells. SEM study also demonstrated the deformed, fragmented, and amorphous nature of BS construing the bactericidal effect of prepared nanoemulsion.
Sigma-Aldrich
1-Octanoyl-rac-glycerol, ≥99%