Merck
  • Characterization of Terpene Synthase from Tea Green Leafhopper Being Involved in Formation of Geraniol in Tea (Camellia sinensis) Leaves and Potential Effect of Geraniol on Insect-Derived Endobacteria.
Characterization of Terpene Synthase from Tea Green Leafhopper Being Involved in Formation of Geraniol in Tea (Camellia sinensis) Leaves and Potential Effect of Geraniol on Insect-Derived Endobacteria.
Biomolecules (2019-12-06)
Ying Zhou, Xiaoyu Liu, Ziyin Yang
ABSTRACT
When insects attack plants, insect-derived elicitors and mechanical damage induce the formation and emission of plant volatiles that have important ecological functions and flavor properties. These events have mainly been studied in model plants, rather than crop plants. Our study showed that tea green leafhopper (Empoasca (Matsumurasca) onukii Matsuda), a major pest infesting tea attack significantly induced the emission of geraniol from tea leaves, but did not affect the crude enzyme activity of geraniol synthase in tea leaves. An enzyme extract of E. (M.) onukii specifically produced geraniol from geraniol diphosphate. Furthermore, a terpene synthase (EoTPS) was isolated from E. (M.) onukii. This terpene synthase was able to convert geraniol diphosphate to geraniol in vitro. In addition, geraniol had in vitro ability to inhibit the growth of Acinetobacter johnsonii that is endobacterial isolated from E. (M.) onukii. This information illustrates that elicitors from piercing-sucking insects can induce the formation of volatiles from crop plants and advances our understanding of the roles of plant volatiles in the interaction among crops-insects-microorganisms.
Sigma-Aldrich
Geraniol, 98%