Merck
  • The role of the SWI/SNF chromatin remodeling complex in maintaining the stemness of glioma initiating cells.
The role of the SWI/SNF chromatin remodeling complex in maintaining the stemness of glioma initiating cells.
Scientific reports (2017-04-20)
Hiroaki Hiramatsu, Kazuyoshi Kobayashi, Kyousuke Kobayashi, Takeshi Haraguchi, Yasushi Ino, Tomoki Todo, Hideo Iba
ABSTRACT
Glioma initiating cells (GICs) are thought to contribute to therapeutic resistance and tumor recurrence in glioblastoma, a lethal primary brain tumor in adults. Although the stem-like properties of GICs, such as self-renewal and tumorigenicity, are epigenetically regulated, the role of a major chromatin remodeling complex in human, the SWI/SNF complex, remains unknown in these cells. We here demonstrate that the SWI/SNF core complex, that is associated with a unique corepressor complex through the d4-family proteins, DPF1 or DPF3a, plays essential roles in stemness maintenance in GICs. The serum-induced differentiation of GICs downregulated the endogenous expression of DPF1 and DPF3a, and the shRNA-mediated knockdown of each gene reduced both sphere-forming ability and tumor-forming activity in a mouse xenograft model. Rescue experiments revealed that DPF1 has dominant effects over DPF3a. Notably, whereas we have previously reported that d4-family members can function as adaptor proteins between the SWI/SNF complex and NF-κB dimers, this does not significantly contribute to maintaining the stemness properties of GICs. Instead, these proteins were found to link a corepressor complex containing the nuclear receptor, TLX, and LSD1/RCOR2 with the SWI/SNF core complex. Collectively, our results indicate that DPF1 and DPF3a are potential therapeutic targets for glioblastoma.
Sigma-Aldrich
Donkey Anti-Rabbit IgG Antibody, HRP conjugate, Species Adsorbed, Chemicon®, from donkey