Merck
  • Evaluation of Ascorbic Acid or Curcumin Formulated in a Solid Dispersion on Salmonella Enteritidis Infection and Intestinal Integrity in Broiler Chickens.
Evaluation of Ascorbic Acid or Curcumin Formulated in a Solid Dispersion on Salmonella Enteritidis Infection and Intestinal Integrity in Broiler Chickens.
Pathogens (Basel, Switzerland) (2019-11-14)
Daniel Hernandez-Patlan, Bruno Solis-Cruz, Karine P Pontin, Juan D Latorre, Xochitl Hernandez-Velasco, Ruben Merino-Guzman, Abraham Mendez-Albores, Billy M Hargis, Raquel Lopez-Arellano, Guillermo Tellez-Isaias
ABSTRACT
Two experimental models were conducted to evaluate and compare the effect of ascorbic acid (AA) or curcumin formulated in a solid dispersion (SD-CUR) as prophylactic or therapeutic alternatives to prevent or control S. Enteritidis (SE) infection in broiler chickens. In the prophylactic model, dietary administration of AA showed a significant reduction in SE counts in crop compared to the positive control (PC) group (p < 0.05), whereas in cecal tonsils (CT), SD-CUR significantly reduced SE recovery. Superoxide dismutase (SOD) activity was significantly higher in chickens supplemented with AA or SD-CUR, and total intestinal IgA levels were significantly lower in both treatments when compared to the PC group. Serum fluorescein isothiocyanate-dextran (FITC-d) levels were reduced by SD-CUR compared to PC, while AA presented significantly lower total aerobic bacteria. In the therapeutic model, only the dietary administration of AA significantly decreased SE in crop and CT on days 3 and 10 post-challenge. FITC-d levels were significantly lower in both treated groups in comparison to PC, but IgA levels were significantly reduced only by AA. The results suggest that dietary AA and SD-CUR have different modes of action to reduce SE intestinal colonization in two different challenge models in broiler chickens.
Product Description
Millipore
Brilliant green agar, modified, suitable for microbiology, NutriSelect® Basic
Sigma-Aldrich
Nalidixic acid sodium salt, powder
Sigma-Aldrich
Novobiocin sodium salt, ≥90% (HPLC)