Merck
  • B7-H3 contributes to the metastatic capacity of melanoma cells by modulation of known metastasis-associated genes.
B7-H3 contributes to the metastatic capacity of melanoma cells by modulation of known metastasis-associated genes.
International journal of cancer (2011-06-15)
Christina Tekle, Marit Kveine Nygren, Yih-Wen Chen, Indrejit Dybsjord, Jahn Marthin Nesland, Gunhild Mari Maelandsmo, Oystein Fodstad
ABSTRACT
B7-H3, an immunoregulatory protein, is known to play a role in tumor progression. In many cancer types, observed correlations between high B7-H3 expression and poor prognosis have been attributed to involvement in antitumor immunity. However, here we demonstrate a nonimmunological alternative function of B7-H3 in cancer metastasis. Since advanced malignant melanoma is a disease with a poor survival rate and a broad pattern of metastasis, we used this disease as a model in our studies. We found that shRNA silencing of B7-H3 reduced the in vitro migratory potential and matrigel invasiveness of MDA-MB-435 and FEMX-I melanoma cells. In an experimental metastasis model in vivo, B7-H3 silencing of MDA-MB-435 cells resulted in reduced metastatic capacity and significantly increased the median symptom-free survival of nude mice (147 vs. 65 days, p < 0.001) and rats (53 vs. 42 days, p = 0.025) injected with MDA-MB-435 cells. Furthermore, a smaller fraction of mice had microscopically detectable metastases compared to control animals, and the pattern of metastases was slightly different between the two groups but with the brain as the predominant organ. Immunohistochemistry on samples from two melanoma patients showed strong B7-H3 staining in both a primary tumor and metastases. Notably, the metastasis-associated proteins, matrix metalloproteinase (MMP)-2, signal transducer and activator of transcription 3 (Stat3), and the level of secreted interleukin-8 (IL-8) were reduced in the B7-H3 knock-down cell variants, whereas tissue inhibitor of metalloproteinase (TIMP)-1 and-2 levels were increased. Taken together, our findings indicate a novel role for B7-H3 in the regulation of the metastatic capacity of melanoma cells and it might be a potential therapeutic target for anti-metastasis therapy.
Sigma-Aldrich
Anti-TIMP-2 Antibody, clone 67-4H11, clone 67-4H11, Chemicon®, from mouse
Sigma-Aldrich
Anti-α-Tubulin Mouse mAb (DM1A), liquid, clone DM1A, Calbiochem®