- Gq-DREADD Selectively Initiates Glial Glutamate Release and Inhibits Cue-induced Cocaine Seeking.
Gq-DREADD Selectively Initiates Glial Glutamate Release and Inhibits Cue-induced Cocaine Seeking.
Glial cells of the central nervous system directly influence neuronal activity by releasing neuroactive small molecules, including glutamate. Long-lasting cocaine-induced reductions in extracellular glutamate in the nucleus accumbens core (NAcore) affect synaptic plasticity responsible for relapse vulnerability. We transduced NAcore astrocytes with an adeno-associated virus vector expressing hM3D designer receptor exclusively activated by a designer drug (DREADD) under control of the glial fibrillary acidic protein promoter in 62 male Sprague Dawley rats, 4 dominant-negative soluble N-ethylmaleimide-sensitive factor attachment protein receptor mice, and 4 wild-type littermates. Using glutamate biosensors, we measured NAcore glutamate levels following intracranial or systemic administration of clozapine N-oxide (CNO) and tested the ability of systemic CNO to inhibit reinstated cocaine or sucrose seeking following self-administration and extinction training. Administration of CNO in glial fibrillary acidic protein-hM3D-DREADD transfected animals increased NAcore extracellular glutamate levels in vivo. The glial origin of released glutamate was validated by an absence of CNO-mediated release in mice expressing a dominant-negative soluble N-ethylmaleimide-sensitive factor attachment protein receptor variant in glia. Also, CNO-mediated release was relatively insensitive to N-type calcium channel blockade. Systemic administration of CNO inhibited cue-induced reinstatement of cocaine seeking in rats extinguished from cocaine but not sucrose self-administration. The capacity to inhibit reinstated cocaine seeking was prevented by systemic administration of the group II metabotropic glutamate receptor antagonist LY341495. DREADD-mediated glutamate gliotransmission inhibited cue-induced reinstatement of cocaine seeking by stimulating release-regulating group II metabotropic glutamate receptor autoreceptors to inhibit cue-induced synaptic glutamate spillover.