Merck
  • Characterization of an atypical gamma-secretase complex from hematopoietic origin.
Characterization of an atypical gamma-secretase complex from hematopoietic origin.
Biochemistry (2010-02-25)
Lisa Placanica, Jennifer W Chien, Yue-Ming Li
ABSTRACT
Gamma-secretase is a widely expressed multisubunit enzyme complex which is involved in the pathogenesis of Alzheimer disease and hematopoietic malignancies through its aberrant processing of the amyloid precursor protein (APP) and Notch1, respectively. While gamma-secretase has been extensively studied, there is a dearth of information surrounding the activity, composition, and function of gamma-secretase expressed in distinct cellular populations. Here we show that endogenous gamma-secretase complexes of hematopoietic origin are distinct from epithelial derived gamma-secretase complexes. Hematopoietic gamma-secretase has reduced activity for APP and Notch1 processing compared to epithelial gamma-secretase. Characterization of the active complexes with small molecule affinity probes reveals that hematopoietic gamma-secretase has an atypical subunit composition with significantly altered subunit stoichiometry. Furthermore, we demonstrate that these discrete complexes exhibit cell-line specific substrate selectivity suggesting a possible mechanism of substrate regulation. These data underscore the need for studying endogenous gamma-secretase to fully understand of the biology of gamma-secretase and its complexity as a molecular target for the development of disease therapeutics.
Sigma-Aldrich
Anti-Presenilin-1 Antibody, loop, a.a. 275-367, CT, serum, Chemicon®
Pricing and availability is not currently available.