Merck
  • Hydrolysis of cellulose derived from steam exploded bagasse by Penicillium cellulases: comparison with commercial cellulase.
Hydrolysis of cellulose derived from steam exploded bagasse by Penicillium cellulases: comparison with commercial cellulase.
Bioresource technology (2009-08-18)
Rajkumar Singh, A J Varma, R Seeta Laxman, Mala Rao
ABSTRACT
A complete cellulase from Penicillium pinophilum was evaluated for the hydrolysis of alpha-cellulose derived from steam exploded sugarcane bagasse and other cellulosic substrates. Alpha-cellulose at 1% substrate concentration was completely hydrolyzed by Penicillium cellulase within 3h wherein at 10% the hydrolysis was 100% within 24 h with an enzyme loading of 10 FPU/g. The hydrolysate yielded glucose as major end product as analyzed by HPLC. Under similar conditions, hydrolysis of Sigmacell (microcrystalline cellulose), CP-123 (pulverized cellulose powder) and ball milled Solka Floc were 42%, 56% and 52%, respectively. Further the hydrolysis performance of Penicillium sp. cellulase is compared with Trichoderma reesei cellulase (Accellerase 1000) from Genencore. The kinetics of hydrolysis with respect to enzyme and substrate concentration will be presented.
Sigma-Aldrich
Sigmacell Cellulose, Type 101, Highly purified, fibers
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm