Merck
Asymmetric total synthesis of antiochic acid.
Organic letters (2008-04-29)
Yu-Jun Zhao, Teck-Peng Loh
ABSTRACT
The first asymmetric total synthesis of antiochic acid using bioinspired polyene cyclization strategy is described. Both good yield and good asymmetric induction were obtained.
Sigma-Aldrich
Isopropenylboronic acid pinacol ester, contains phenothiazine as stabilizer, 95%