Merck
  • Selenoprotein Transcript Level and Enzyme Activity as Biomarkers for Selenium Status and Selenium Requirements in the Turkey (Meleagris gallopavo).
Selenoprotein Transcript Level and Enzyme Activity as Biomarkers for Selenium Status and Selenium Requirements in the Turkey (Meleagris gallopavo).
PloS one (2016-03-24)
Rachel M Taylor, Roger A Sunde
ABSTRACT
The current National Research Council (NRC) selenium (Se) requirement for the turkey is 0.2 μg Se/g diet. The sequencing of the turkey selenoproteome offers additional molecular biomarkers for assessment of Se status. To determine dietary Se requirements using selenoprotein transcript levels and enzyme activities, day-old male turkey poults were fed a Se-deficient diet supplemented with graded levels of Se (0, 0.025, 0.05, 0.1, 0.2, 0.3, 0.4, 0.5, 0.75, 1.0 μg Se/g diet) as selenite, and 12.5X the vitamin E requirement. Poults fed less than 0.05 μg Se/g diet had a significantly reduced rate of growth, indicating the Se requirement for growth in young male poults is 0.05 μg Se/g diet. Se deficiency decreased plasma GPX3 (glutathione peroxidase), liver GPX1, and liver GPX4 activities to 2, 3, and 7%, respectively, of Se-adequate levels. Increasing Se supplementation resulted in well-defined plateaus for all blood, liver and gizzard enzyme activities and mRNA levels, showing that these selenoprotein biomarkers could not be used as biomarkers for supernutritional-Se status. Using selenoenzyme activity, minimum Se requirements based on red blood cell GPX1, plasma GPX3, and pancreas and liver GPX1 activities were 0.29-0.33 μg Se/g diet. qPCR analyses using all 10 dietary Se treatments for all 24 selenoprotein transcripts (plus SEPHS1) in liver, gizzard, and pancreas found that only 4, 4, and 3 transcripts, respectively, were significantly down-regulated by Se deficiency and could be used as Se biomarkers. Only GPX3 and SELH mRNA were down regulated in all 3 tissues. For these transcripts, minimum Se requirements were 0.07-0.09 μg Se/g for liver, 0.06-0.15 μg Se/g for gizzard, and 0.13-0.18 μg Se/g for pancreas, all less than enzyme-based requirements. Panels based on multiple Se-regulated transcripts were effective in identifying Se deficiency. These results show that the NRC turkey dietary Se requirement should be raised to 0.3 μg Se/g diet.
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
DL-α-Tocopherol acetate, ≥96% (HPLC)