Merck
  • Adsorption and degradation of imazapic in soils under different environmental conditions.
Adsorption and degradation of imazapic in soils under different environmental conditions.
PloS one (2019-07-10)
Wangcang Su, Hongdan Hao, Mingzhen Ding, Renhai Wu, Hongle Xu, Fei Xue, Changchao Shen, Lanlan Sun, Chuantao Lu
ABSTRACT
Imazapic is widely used in peanut production, and its residues can cause damage to succeeding crops planted in the following year. The planting area of peanut is large in Henan province. Inceptisol is the main soil type in Henan Province and was used in laboratory experiments that were conducted to investigate imazapic degradation in soil under various environmental conditions. The results indicated that the imazapic degradation rate increased with an increase in temperature, soil pH, and soil moisture, and decreased with organic matter content. The use of biogas slurry as a soil amendment accelerated imazapic degradation. The half-life of imazapic in sterilized soil (364.7 d) was longer than in unsterilized soil (138.6 d), which suggested that there was a significant microbial contribution to imazapic degradation. Imazapic adsorption was also examined and was found to be well described by the Freundlich isotherm. The results indicate that soil has a certain adsorption capacity for imazapic.
Product Description
Supelco
Imazapic, PESTANAL®, analytical standard