Merck
  • Cancer Targeted Gene Therapy for Inhibition of Melanoma Lung Metastasis with eIF3i shRNA Loaded Liposomes.
Cancer Targeted Gene Therapy for Inhibition of Melanoma Lung Metastasis with eIF3i shRNA Loaded Liposomes.
Molecular pharmaceutics (2019-11-26)
Wen Xiao, Weiyi Zhang, Hai Huang, Yafei Xie, Yi Zhang, Xia Guo, Chaohui Jin, Xuelian Liao, Shaohua Yao, Guo Chen, Xiangrong Song
ABSTRACT
Eukaryotic translation initiation factors 3i (eIF3i) is a proto-oncogene that is overexpressed in various tumors, reducing its expression by eIF3i shRNA is a promising strategy to inhibit tumor growth or metastasis. Tumor cell is the target of eIF3i shRNA so that tumor-site accumulation could be important for fulfilling its therapeutic effect. Thus, the iRGD modified liposome (R-LP) was rationally synthesized to enhance the antitumor effect by active targeted delivery of eIF3i shRNA to B16F10 melanoma cells. R-LP encapsulating eIF3i shRNA gene (R-LP/sheIF3i) were prepared by a film dispersion method. The transfection experiment proves that R-LP could effectively transfect B16F10 cells. R-LP/sheIF3i notably restrained the migration, invasion, and adhesion of melanoma cells in vitro. In a mouse model of lung metastasis, R-LP/sheIF3i administered by intravenous injection suppressed pulmonary metastasis of melanoma by dramatically downregulated eIF3i expression and subsequently inhibiting tumor neovascularization and tumor cells proliferation in vivo. Our results provide a basis for tumor cells targeting strategies to reduce the expression of eIF3i by RNAi in the treatment of tumor metastasis.