Merck
  • Breast cancer cells rely on environmental pyruvate to shape the metastatic niche.
Breast cancer cells rely on environmental pyruvate to shape the metastatic niche.
Nature (2019-03-01)
Ilaria Elia, Matteo Rossi, Steve Stegen, Dorien Broekaert, Ginevra Doglioni, Marit van Gorsel, Ruben Boon, Carmen Escalona-Noguero, Sophie Torrekens, Catherine Verfaillie, Erik Verbeken, Geert Carmeliet, Sarah-Maria Fendt
ABSTRACT
The extracellular matrix is a major component of the local environment-that is, the niche-that determines cell behaviour1. During metastatic growth, cancer cells shape the extracellular matrix of the metastatic niche by hydroxylating collagen to promote their own metastatic growth2,3. However, only particular nutrients might support the ability of cancer cells to hydroxylate collagen, because nutrients dictate which enzymatic reactions are active in cancer cells4,5. Here we show that breast cancer cells rely on the nutrient pyruvate to drive collagen-based remodelling of the extracellular matrix in the lung metastatic niche. Specifically, we discovered that pyruvate uptake induces the production of α-ketoglutarate. This metabolite in turn activates collagen hydroxylation by increasing the activity of the enzyme collagen prolyl-4-hydroxylase (P4HA). Inhibition of pyruvate metabolism was sufficient to impair collagen hydroxylation and consequently the growth of breast-cancer-derived lung metastases in different mouse models. In summary, we provide a mechanistic understanding of the link between collagen remodelling and the nutrient environment in the metastatic niche.
Product Description
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in a glass vial
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid