- The Inhibition by Guanfu Base A of Neuropathic Pain Mediated by P2Y12 Receptor in Dorsal Root Ganglia.
The Inhibition by Guanfu Base A of Neuropathic Pain Mediated by P2Y12 Receptor in Dorsal Root Ganglia.
Activation of satellite glial cells (SGCs) in the dorsal root ganglia (DRG) is involved in mechanical and thermal hyperalgesia. The upregulated P2Y12 receptor expressed in SGCs of the DRG participates in the nociceptive transmission of neuropathic pain. Guanfu base A (GFA) has been reported to exhibit antiarrhythmic and anti-inflammatory effects. In this study, we explored the effects of GFA on P2Y12 receptor-mediated mechanical and thermal hyperalgesia in chronic constriction injury (CCI) rats. Sprague-Dawley rats were randomly divided into sham operation group (Sham), CCI operation group (CCI), CCI rats treated with guanfu base A group (CCI + GFA) and control rats treated with GFA group (Ctrl + GFA). Mechanical withdrawal threshold and thermal withdrawal latency were measured. P2Y12 expression in L4-L6 dorsal root ganglion (DRG) was detected by quantitative real-time PCR and Western blot. After CCI treatment, mechanical and thermal hyperalgesia and the expression values of P2Y12 receptor mRNA and protein in DRG were increased. Dual-labeling immunofluorescence showed that the coexpression of P2Y12 receptor and glial fibrillary acidic protein (GFAP) in the DRG of CCI rats was increased compared to sham rats. GFA relieved mechanical and thermal hyperalgesia in the CCI rats, decreased the expression of P2Y12 mRNA and protein and phosphorylation of p38 MAPK in the DRG, and increased the ADP-downregulated cAMP concentrations in HEK293 cells transfected with P2Y12 plasmid. After CCI rats were treated with GFA, the coexpression of P2Y12 receptor and GFAP in the DRG was significantly decreased compared to the untreated CCI group. Thus, downregulating the P2Y12 receptor relieved mechanical and thermal hyperalgesia in the CCI rats.