- Sphingosine 1-phosphate-mediated activation of ezrin-radixin-moesin proteins contributes to cytoskeletal remodeling and changes of membrane properties in epithelial otic vesicle progenitors.
Sphingosine 1-phosphate-mediated activation of ezrin-radixin-moesin proteins contributes to cytoskeletal remodeling and changes of membrane properties in epithelial otic vesicle progenitors.
Hearing loss is among the most prevalent sensory impairments in humans. Cochlear implantable devices represent the current therapies for hearing loss but have various shortcomings. ERM (ezrin- radixin -moesin) are a family of adaptor proteins that link plasma membrane with actin cytoskeleton, playing a crucial role in cell morphology and in the formation of membrane protrusions. Recently, bioactive sphingolipids have emerged as regulators of ERM proteins. Sphingosine 1-phosphate (S1P) is a pleiotropic sphingolipid which regulates fundamental cellular functions such as proliferation, survival, migration as well as processes such as development and inflammation mainly via ligation to its specific receptors S1PR (S1P1-5). Experimental findings demonstrate a key role for S1P signaling axis in the maintenance of auditory function. Preservation of cellular junctions is a fundamental function both for S1P and ERM proteins, crucial for the maintenance of cochlear integrity. In the present work, S1P was found to activate ERM in a S1P2-dependent manner in murine auditory epithelial progenitors US/VOT-E36. S1P-induced ERM activation potently contributed to actin cytoskeletal remodeling and to the appearance of ionic currents and membrane passive properties changes typical of more differentiated cells. Moreover, PKC and Akt activation was found to mediate S1P-induced ERM phosphorylation. The obtained findings contribute to demonstrate the role of S1P signaling pathway in inner ear biology and to disclose potential innovative therapeutical approaches in the field of hearing loss prevention and treatment.