Merck
  • Ambient Air Pollution Is Associated With HDL (High-Density Lipoprotein) Dysfunction in Healthy Adults.
Ambient Air Pollution Is Associated With HDL (High-Density Lipoprotein) Dysfunction in Healthy Adults.
Arteriosclerosis, thrombosis, and vascular biology (2019-02-01)
Jianping Li, Changping Zhou, Hongbing Xu, Robert D Brook, Shengcong Liu, Tieci Yi, Yang Wang, Baihuan Feng, Mingming Zhao, Xu Wang, Qian Zhao, Jie Chen, Xiaoming Song, Tong Wang, Shuo Liu, Yi Zhang, Rongshan Wu, Jianing Gao, Bing Pan, Subramaniam Pennathur, Sanjay Rajagopalan, Yong Huo, Lemin Zheng, Wei Huang
ABSTRACT
Objective- We aimed to assess whether exposure to higher levels of ambient air pollution impairs HDL (high-density lipoprotein) function and to elucidate the underlying biological mechanisms potentially involved. Approach and Results- In the Beijing AIRCHD study (Air Pollution and Cardiovascular Dysfunction in Healthy Adults), 73 healthy adults (23.3±5.4 years) were followed-up with 4 repeated study visits in 2014 to 2016. During each visit, ambient air pollution concentrations, HDL function metrics, and parameters of inflammation and oxidative stress were measured. Average daily concentrations of ambient particulate matter in diameter <2.5 μm were 62.9 µg/m3 (8.1-331.0 µg/m3). We observed significant decreases in HDL cholesterol efflux capacity of 2.3% (95% CI, -4.3 to -0.3) to 5.0% (95% CI, -7.6 to -2.4) associated with interquartile range increases in moving average concentrations of particulate matter in diameter <2.5 μm and traffic-related air pollutants (black carbon, nitrogen dioxide, and carbon monoxide) during the 1 to 7 days before each participant's clinic visit. Higher ambient air pollutant levels were also associated with significant reductions in circulating HDL cholesterol and apoA-I (apolipoprotein A-I), as well as elevations in HDL oxidation index, oxidized LDL (low-density lipoprotein), malondialdehyde, and high-sensitivity C-reactive protein. Conclusions- Higher ambient air pollution concentrations were associated with impairments in HDL functionality, potentially because of systemic inflammation and oxidative stress. These novel findings further our understanding of the mechanisms whereby air pollutants promote cardiometabolic disorders.
Sigma-Aldrich
Dextran sulfate sodium salt from Leuconostoc spp., Molecular Biology, average Mw >500,000 (dextran starting material), contains 0.5-2% phosphate buffer