Merck
  • Triboionization: a Novel Ionization Method by Peeling of Cohesive Substances for Mass Spectrometry.
Triboionization: a Novel Ionization Method by Peeling of Cohesive Substances for Mass Spectrometry.
Journal of the American Society for Mass Spectrometry (2019-05-19)
Natsuhiko Sugimura, Yuki Watabe, Toshimichi Shibue
ABSTRACT
A novel ionization/sampling method termed triboionization was developed. Triboionization is an ionization method that only uses cohesive substances, such as food wrap or sticky tape, and does not require an electrode, electric power supply, heat source, light source, radiation, or gas, unlike most other conventional ambient ionization methods. In this study, the sample compound attached to adhesive tape or plastic wrap was quickly peeled off at a distance of approximately 2 cm from the atmospheric interface of a mass spectrometer. All of the five types of food wrap and 13 types of adhesive tape tested successfully ionized caffeine. Nine out of ten model compounds were detected as the corresponding molecular ions in the positive or negative mode by this ionizing contrivance using an oriented polypropylene adhesive tape. The detected molecular ions were typically protonated molecules or sodium adducts in the positive mode or deprotonated molecules in the negative mode. The elemental compositions of the observed ions were confirmed within 5 ppm by high-resolution mass spectrometry. The triboionization phenomenon was considered to depend on physical and electronic events caused by peeling off a cohesive substance. Triboionization is able to provide a compact ion source using only mechanical mechanisms. Additionally, triboionization allows sticky tape to be used as a convenient sampling device for surface analysis.