Merck
Capturing the interactome of newly transcribed RNA.
Nature methods (2018-02-13)
Xichen Bao, Xiangpeng Guo, Menghui Yin, Muqddas Tariq, Yiwei Lai, Shahzina Kanwal, Jiajian Zhou, Na Li, Yuan Lv, Carlos Pulido-Quetglas, Xiwei Wang, Lu Ji, Muhammad J Khan, Xihua Zhu, Zhiwei Luo, Changwei Shao, Do-Hwan Lim, Xiao Liu, Nan Li, Wei Wang, Minghui He, Yu-Lin Liu, Carl Ward, Tong Wang, Gong Zhang, Dongye Wang, Jianhua Yang, Yiwen Chen, Chaolin Zhang, Ralf Jauch, Yun-Gui Yang, Yangming Wang, Baoming Qin, Minna-Liisa Anko, Andrew P Hutchins, Hao Sun, Huating Wang, Xiang-Dong Fu, Biliang Zhang, Miguel A Esteban
ABSTRACT
We combine the labeling of newly transcribed RNAs with 5-ethynyluridine with the characterization of bound proteins. This approach, named capture of the newly transcribed RNA interactome using click chemistry (RICK), systematically captures proteins bound to a wide range of RNAs, including nascent RNAs and traditionally neglected nonpolyadenylated RNAs. RICK has identified mitotic regulators amongst other novel RNA-binding proteins with preferential affinity for nonpolyadenylated RNAs, revealed a link between metabolic enzymes/factors and nascent RNAs, and expanded the known RNA-bound proteome of mouse embryonic stem cells. RICK will facilitate an in-depth interrogation of the total RNA-bound proteome in different cells and systems.
Sigma-Aldrich
5-Ethynyl uridine, ≥95%