Merck
  • Effects of Bt Corn on the Development and Fecundity of Corn Earworm (Lepidoptera: Noctuidae).
Effects of Bt Corn on the Development and Fecundity of Corn Earworm (Lepidoptera: Noctuidae).
Journal of economic entomology (2018-07-10)
Tom R Bilbo, Francis P F Reay-Jones, Dominic D Reisig, Fred R Musser, Jeremy K Greene
ABSTRACT
The corn earworm, Helicoverpa zea (Boddie) (Lepidoptera: Noctuidae), is only moderately susceptible to most toxins from the bacterium Bacillus thuringiensis (Bt) expressed in transgenic corn. To better understand the impact of Bt corn on the life cycle of H. zea, we collected pupae of H. zea during 2014-2016 in North Carolina, South Carolina, and Mississippi from corn hybrids expressing Cry1Ab, Cry1F, Cry1F + Cry1Ab, Cry1F + Cry1Ab + Vip3Aa20, Cry1A.105 + Cry2Ab2, and Cry1A.105 + Cry2Ab2 + Cry1F, as well as from non-Bt near-isolines. We investigated the effect of Bt corn on pupal weight, fecundity, and egg viability of H. zea. Pupal weights were significantly reduced for males and females from all Bt hybrids compared with non-Bt near-isolines. Female pupae from the hybrid expressing Cry1F + Cry1Ab were also significantly lighter relative to those from the near-isolines expressing only Cry1F. Reductions in pupal weight did not result in any detectable effects on fecundity or egg viability. The reduction in pupal weight in the hybrids expressing Cry1F and Cry1F + Cry1Ab significantly declined over time in South Carolina, possibly indicating developing resistance to these Bt toxins. These data can be incorporated into insect resistance management models used to improve risk management decisions regarding H. zea in Bt crops in the complex landscapes of the southern United States.
Product Description
Supelco
Hydramethylnon, PESTANAL®, analytical standard