Merck
  • ER-mitochondria cross-talk is regulated by the Ca2+ sensor NCS1 and is impaired in Wolfram syndrome.
ER-mitochondria cross-talk is regulated by the Ca2+ sensor NCS1 and is impaired in Wolfram syndrome.
Science signaling (2018-10-26)
Claire Angebault, Jérémy Fauconnier, Simone Patergnani, Jennifer Rieusset, Alberto Danese, Corentin A Affortit, Jolanta Jagodzinska, Camille Mégy, Mélanie Quiles, Chantal Cazevieille, Julia Korchagina, Delphine Bonnet-Wersinger, Dan Milea, Christian Hamel, Paolo Pinton, Marc Thiry, Alain Lacampagne, Benjamin Delprat, Cécile Delettre
ABSTRACT
Communication between the endoplasmic reticulum (ER) and mitochondria plays a pivotal role in Ca2+ signaling, energy metabolism, and cell survival. Dysfunction in this cross-talk leads to metabolic and neurodegenerative diseases. Wolfram syndrome is a fatal neurodegenerative disease caused by mutations in the ER-resident protein WFS1. Here, we showed that WFS1 formed a complex with neuronal calcium sensor 1 (NCS1) and inositol 1,4,5-trisphosphate receptor (IP3R) to promote Ca2+ transfer between the ER and mitochondria. In addition, we found that NCS1 abundance was reduced in WFS1-null patient fibroblasts, which showed reduced ER-mitochondria interactions and Ca2+ exchange. Moreover, in WFS1-deficient cells, NCS1 overexpression not only restored ER-mitochondria interactions and Ca2+ transfer but also rescued mitochondrial dysfunction. Our results describe a key role of NCS1 in ER-mitochondria cross-talk, uncover a pathogenic mechanism for Wolfram syndrome, and potentially reveal insights into the pathogenesis of other neurodegenerative diseases.
Sigma-Aldrich
MISSION® esiRNA, targeting human WFS1
Sigma-Aldrich
Tetramethylrhodamine methyl ester perchlorate, ≥95%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
JC-1, powder or solid (Crystals)