Merck
  • Modeling Wnt signaling by CRISPR-Cas9 genome editing recapitulates neoplasia in human Barrett epithelial organoids.
Modeling Wnt signaling by CRISPR-Cas9 genome editing recapitulates neoplasia in human Barrett epithelial organoids.
Cancer letters (2018-08-26)
Xi Liu, Yulan Cheng, John M Abraham, Zhixiong Wang, Zhe Wang, Xiquan Ke, Rong Yan, Eun Ji Shin, Saowanee Ngamruengphong, Mouen A Khashab, Guanjun Zhang, George McNamara, Andrew J Ewald, DeChen Lin, Zhengwen Liu, Stephen J Meltzer
ABSTRACT
Primary organoid cultures generated from patient biopsies comprise a novel improved platform for disease modeling, being genetically stable and closely recapitulating in vivo scenarios. Barrett esophagus (BE) is the major risk factor for esophageal adenocarcinoma. There has been a dearth of long-term in vitro expansion models of BE neoplastic transformation. We generated a long-term virus-free organoid expansion model of BE neoplasia from patient biopsies. Both wild-type and paired APC-knockout (APCKO) BE organoids genome-edited by CRISPR-Cas9 showed characteristic goblet cell differentiation. Autonomous Wnt activation was confirmed in APCKO organoids by overexpression of Wnt target genes and nuclear-translocated β-catenin expression after withdrawal of Wnt-3A and R-spondin-1. Wnt-activated organoids demonstrated histologic atypia, higher proliferative and replicative activity, reduced apoptosis, and prolonged culturability. Wnt-activated organoids also showed sustained protrusive migration ability accompanied by disrupted basement membrane reorganization and integrity. This CRISPR-Cas9 editing human-derived organoid model recapitulates the critical role of aberrant Wnt/β-catenin signaling activation in BE neoplastic transformation. This system can be used to study other 'driver' pathway alterations in BE-associated neoplasia, avoiding signaling noise present in immortalized or cancer-derived cell lines.
Sigma-Aldrich
Nuclear Fast Red
Sigma-Aldrich
Anti-Collagen Type IV Antibody, Chemicon®, from goat