Merck
  • Quantitative analysis of phospholipids using nanostructured laser desorption ionization targets.
Quantitative analysis of phospholipids using nanostructured laser desorption ionization targets.
Lipids (2011-02-18)
Simona Colantonio, John T Simpson, Robert J Fisher, Amichai Yavlovich, Julie M Belanger, Anu Puri, Robert Blumenthal
ABSTRACT
Since its introduction as an ionization technique in mass spectrometry, matrix-assisted laser desorption ionization (MALDI) has been applied to a wide range of applications. Quantitative small molecule analysis by MALDI, however, is limited due to the presence of intense signals from the matrix coupled with non-homogeneous surfaces. The surface used in nano-structured laser desorption ionization (NALDI) eliminates the need for a matrix and the resulting interferences, and allows for quantitative analysis of small molecules. This study was designed to analyze and quantitate phospholipid components of liposomes. Here we have developed an assay to quantitate the DPPC and DC(8,9)PC in liposomes by NALDI following various treatments. To test our method we chose to analyze a liposome system composed of DPPC (1,2-dipalmitoyl-sn-glycero-3-phosphocholine) and DC(8,9)PC (1,2-bis(tricosa-10,12-diynoyl)-sn-glycero-3-phosphocholine), as DC(8,9)PC is known to undergo cross-linking upon treatment with UV (254 nm) and this reaction converts the monomer into a polymer. First, calibration curves for pure lipids (DPPC and DC(8,9)PC) were created using DMPC (1,2-dimyristoyl-sn-glycero-3-phosphocholine) as an internal standard. The calibration curve for both DPPC and DC(8,9)PC showed an R(2) of 0.992, obtained using the intensity ratio of analyte and internal standard. Next, DPPC:DC(8,9)PC liposomes were treated with UV radiation (254 nm). Following this treatment, lipids were extracted from the liposomes and analyzed. The analysis of the lipids before and after UV exposure confirmed a decrease in the signal of DC(8,9)PC of about 90%. In contrast, there was no reduction in DPPC signal.