Merck
  • Cloning and characterization of the human PIM-1 gene: a putative oncogene related to the protein kinases.
Cloning and characterization of the human PIM-1 gene: a putative oncogene related to the protein kinases.
Journal of cellular biochemistry (1987-10-01)
T C Meeker, L Nagarajan, A ar-Rushdi, C M Croce
ABSTRACT
The mouse PIM-1 gene has been implicated in the evolution of retrovirus-associated mouse lymphomas. We have initiated a study of the human PIM-1 gene because of its potential importance as a human oncogene. We have isolated genomic and cDNA clones for this gene and characterized this locus in detail. The predicted PIM-1 protein is 313 amino acids in length. It has homology to a number of the protein kinases but does not have a transmembrane region. The amino acid corresponding to tyrosine-416 of pp60v-src is a tyrosine (position 198), which is consistent with the hypothesis that PIM-1 is a tyrosine kinase rather than a serine-threonine kinase. The PIM-1 gene was found to have six exons and five introns derived from 5 kb of genomic DNA. The site of transcription initiation was localized by S1 nuclease protection studies which indicated that the mature PIM-1 mRNA was approximately 2.7 kb in length. The promotor of this gene had no TATA or CAAT box but did have multiple GC boxes (CCGCCC) that might bind the Sp1 protein. The PIM-1 gene was expressed in myeloid and B lymphoid cell lines, but not in T lymphoid and nonhemopoietic lines. This initial characterization of PIM-1 will allow us to define its role in normal and malignant hematolymphoid differentiation.
Sigma-Aldrich
PIM1, active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution