Merck
  • Effects of the chromanol HMR 1556 on potassium currents in atrial myocytes.
Effects of the chromanol HMR 1556 on potassium currents in atrial myocytes.
Naunyn-Schmiedeberg's archives of pharmacology (2003-03-20)
Ralph F Bosch, Alexander C Schneck, Saskia Csillag, Bernd Eigenberger, Uwe Gerlach, Joachim Brendel, Hans J Lang, Christian Mewis, Heinz Gögelein, Ludger Seipel, Volker Kühlkamp
ABSTRACT
The chromanol HMR 1556 is a potent blocker of KvLQT1/minK potassium channels expressed in Xenopus oocytes. The compound is therefore a new class III antiarrhythmic drug with a distinct mechanism of action. However, the effect of HMR 1556 on atrial ion channels and the selectivity of block in the human heart has not been investigated. We tested the effects of HMR 1556 on repolarizing potassium currents in human and guinea pig atrial myocytes. Single atrial myocytes were isolated by enzymatic dissociation. Atrial potassium currents (I(Ks), I(Kr), in guinea pig, I(to), I(Kur), I(K1) in humans) were recorded at 36 degrees C in the whole cell mode of the patch clamp technique. HMR 1556 produced a concentration-dependent and reversible block of I(Ks) with a half maximal concentration (EC(50)) of 6.8 nmol/l. 10 micromol/l HMR 1556 almost completely inhibited I(Ks) (97.2+/-3.2%, n=6). Steady-state activation as well as kinetic properties of the current were not altered by HMR 1556. I(Kr) currents were not affected up to concentrations of 10 micromol/l. HMR 1556 did not inhibit other potassium currents in human atrium: I(to), I(Kur) and the classical inward rectifier potassium current I(K1) were not significantly affected up to concentrations that completely blocked I(Ks) (10 micromol/l). HMR 1556 is a highly-potent blocker of I(Ks) channels without exerting effects on other potassium currents involved in atrial repolarization. Given the potential advantages of I(Ks) vs. I(Kr) blockade, the drug's new mechanism of action warrants further investigation to clarify its role as an antiarrhythmic agent.
Sigma-Aldrich
HMR-1556, ≥98% (HPLC)