Merck
  • Luminescence of Eu3+ ions in hybrid polymer-inorganic composites based on poly(methyl methacrylate) and zirconia nanoparticles.
Luminescence of Eu3+ ions in hybrid polymer-inorganic composites based on poly(methyl methacrylate) and zirconia nanoparticles.
Luminescence : the journal of biological and chemical luminescence (2018-04-24)
Alexander N Bugrov, Anastasia Yu Zavialova, Ruslan Yu Smyslov, Tatyana D Anan'eva, Elena N Vlasova, Maxim V Mokeev, Artem E Kryukov, Gennady P Kopitsa, Vitaliy Pipich
ABSTRACT
Spherical nanoparticles of ZrO2 with 2 and 10 mol% EuO1.5 up to 20 nm size were prepared by the method of hydrothermal synthesis for luminescent functionalization of the polymer-inorganic nanocomposites based on poly(methyl methacrylate). Surface modification of oxide nanoparticles was carried out by 3-(trimethoxysilyl)propyl methacrylate, dimethoxymethylvinyl silane and 2-hydroxyethyl methacrylate to provide uniform distribution and to prevent agglomeration of nanosized filler in the polymer matrix. Polymer-inorganic composites were synthesized by in situ free radical polymerization in bulk. Structuring of ZrO2 -EuO1.5 nanoparticles in the poly(methyl methacrylate) was studied by very-small-angle neutron scattering. According to the results, the dependence of photoluminescent properties of ZrO2 -EuO1.5 nanoparticles on the content of lanthanide, the symmetry of the crystal field, surface treatment and the polymer matrix were established. A correlation was shown between Stark splitting in luminescence spectra of ZrO2 -EuO1.5 nanoparticles and their phase composition. Using MMT-assay it was shown that composites based on poly(methyl methacrylate) and ZrO2 -EuO1.5 nanoparticles do not have cytotoxic properties, which makes it possible to use them as prosthesis materials with contrasted and luminescent imaging properties.
Sigma-Aldrich
Dimethoxymethylvinylsilane, 97%