Merck
Adsorption of terpenic compounds onto organo-palygorskite.
Environmental science and pollution research international (2017-05-14)
Sana Ghrab, Mabrouk Eloussaief, Stéphanie Lambert, Samir Bouaziz, Mourad Benzina
ABSTRACT
Essential oils from aromatic plants are currently mentioned as suitable tools for excellent protection of stored grains from insect pest attacks. The present work aimed to study the processes of the synthesis insecticidal formulation with clay. The active terpenic compounds of essential oil of Eucalyptus globulus (Eg) were fixed in the palygorskite by adsorption process. Two sample types of palygorskites were used: raw and organo-palygorskite. The palygorskite clays were characterized by different physicochemical techniques including X-ray diffraction (XRD), Fourier transform infrared (FTIR) analyses, thermogravimetric analysis (TG), differential thermal analysis (DTA), Brunauer-Emmet-Teller (BET), and scanning electron microscope (SEM). Results reveal that the raw clay has a fibrous structure with impurities essential calcite. These structures and physicochemical properties of raw palygorskite and organo-palygorskite give it the potential of material adsorbent. Results show that the adsorption capacity strongly depends on affinity between terpenic compounds and organic cations rather than on interlayer distance of organo-palygorskite. The highest adsorption capacity of terpenic compounds is acquired with palygorskite interlaced by didodecyldimethylammonium bromide (DDDMA). These results validated the potential utility of the Paly-DDDMA as adsorbent fibrous clay for the retention of terpenic compounds in application of environmental preservation.
Sigma-Aldrich
Didodecyldimethylammonium bromide, 98%