Merck
  • Oxidized phosphatidylcholines are produced in renal ischemia reperfusion injury.
Oxidized phosphatidylcholines are produced in renal ischemia reperfusion injury.
PloS one (2018-04-24)
Zahra Solati, Andrea L Edel, Yue Shang, Karmin O, Amir Ravandi
ABSTRACT
The aim of this study was to determine the individual oxidized phosphatidylcholine (OxPC) molecules generated during renal ischemia/ reperfusion (I/R) injury. Kidney ischemia was induced in male Sprague-Dawley rats by clamping the left renal pedicle for 45 min followed by reperfusion for either 6h or 24h. Kidney tissue was subjected to lipid extraction. Phospholipids and OxPC species were identified and quantitated using liquid chromatography coupled to electrospray ionization tandem mass spectrometry using internal standards. We identified fifty-five distinct OxPC in rat kidney following I/R injury. These included a variety of fragmented (aldehyde and carboxylic acid containing species) and non-fragmented products. 1-stearoyl-2-linoleoyl-phosphatidylcholine (SLPC-OH), which is a non-fragmented OxPC and 1-palmitoyl-2-azelaoyl-sn-glycero-3-phosphocholine (PAzPC), which is a fragmented OxPC, were the most abundant OxPC species after 6h and 24 h I/R respectively. Total fragmented aldehyde OxPC were significantly higher in 6h and 24h I/R groups compared to sham operated groups (P = 0.03, 0.001 respectively). Moreover, levels of aldehyde OxPC at 24h I/R were significantly greater than those in 6h I/R (P = 0.007). Fragmented carboxylic acid increased significantly in 24h I/R group compared with sham and 6h I/R groups (P = 0.001, 0.001). Moreover, levels of fragmented OxPC were significantly correlated with creatinine levels (r = 0.885, P = 0.001). Among non-fragmented OxPC, only isoprostanes were elevated significantly in 6h I/R group compared with sham group but not in 24h I/R group (P = 0.01). No significant changes were observed in other non-fragmented OxPC including long chain products and terminal furans. We have shown for the first time that bioactive OxPC species are produced in renal I/R and their levels increase with increasing time of reperfusion in a kidney model of I/R and correlate with severity of I/R injury. Given the pathological activity of fragmented OxPCs, therapies focused on their reduction may be a mechanism to attenuate renal I/R injury.