Merck
  • CORM-401 induces calcium signalling, NO increase and activation of pentose phosphate pathway in endothelial cells.
CORM-401 induces calcium signalling, NO increase and activation of pentose phosphate pathway in endothelial cells.
The FEBS journal (2018-02-22)
Patrycja Kaczara, Bartosz Proniewski, Christopher Lovejoy, Kamil Kus, Roberto Motterlini, Andrey Y Abramov, Stefan Chlopicki
ABSTRACT
Carbon monoxide-releasing molecules (CO-RMs) induce nitric oxide (NO) release (which requires NADPH), and Ca2+ -dependent signalling; however, their contribution in mediating endothelial responses to CO-RMs is not clear. Here, we studied the effects of CO liberated from CORM-401 on NO production, calcium signalling and pentose phosphate pathway (PPP) activity in human endothelial cell line (EA.hy926). CORM-401 induced NO production and two types of calcium signalling: a peak-like calcium signal and a gradual increase in cytosolic calcium. CORM-401-induced peak-like calcium signal, originating from endoplasmic reticulum, was reduced by thapsigargin, a SERCA inhibitor, and by dantrolene, a ryanodine receptors (RyR) inhibitor. In contrast, the phospholipase C inhibitor U73122 did not significantly affect peak-like calcium signalling, but a slow and progressive CORM-401-induced increase in cytosolic calcium was dependent on store-operated calcium entrance. CORM-401 augmented coupling of endoplasmic reticulum and plasmalemmal store-operated calcium channels. Interestingly, in the presence of NO synthase inhibitor (l-NAME) CORM-401-induced increases in NO and cytosolic calcium were both abrogated. CORM-401-induced calcium signalling was also inhibited by superoxide dismutase (poly(ethylene glycol)-SOD). Furthermore, CORM-401 accelerated PPP, increased NADPH concentration and decreased the ratio of reduced to oxidized glutathione (GSH/GSSG). Importantly, CORM-401-induced NO increase was inhibited by the PPP inhibitor 6-aminonicotinamide (6-AN), but neither by dantrolene nor by an inhibitor of large-conductance calcium-regulated potassium ion channel (paxilline). The results identify the primary role of CO-induced NO increase in the regulation of endothelial calcium signalling, that may have important consequences in controlling endothelial function.
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
6-Aminonicotinamide, 99%
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder