Merck
Stir bar sorptive extraction for trace analysis.
Journal of chromatography. A (2007-01-24)
Frank David, Pat Sandra
ABSTRACT
Stir bar sorptive extraction (SBSE) was introduced in 1999 as a solventless sample preparation method for the extraction and enrichment of organic compounds from aqueous matrices. The method is based on sorptive extraction, whereby the solutes are extracted into a polymer coating on a magnetic stirring rod. The extraction is controlled by the partitioning coefficient of the solutes between the polymer coating and the sample matrix and by the phase ratio between the polymer coating and the sample volume. For a polydimethylsiloxane coating and aqueous samples, this partitioning coefficient resembles the octanol-water partitioning coefficient. In comparison to solid phase micro-extraction, a larger amount of sorptive extraction phase is used and consequently extremely high sensitivities can be obtained as illustrated by several successful applications in trace analysis in environmental, food and biomedical fields. Initially SBSE was mostly used for the extraction of compounds from aqueous matrices. The technique has also been applied in headspace mode for liquid and solid samples and in passive air sampling mode. In this review article, the principles of stir bar sorptive extraction are described and an overview of SBSE applications is given.
Product Description
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with manual holder, needle size 23 ga
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 23 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with autosampler
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 23 ga, PDMS/DVB StableFlex, for use with autosampler
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with manual holder or autosampler, fiber L 2 cm
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME Portable Field Sampler, coating PDMS/DVB
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex, for use with autosampler
Supelco
SPME Sampling Stand, for use with 4 mL vials
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), fused silica fiber, df 65 μm(PDMS/DVB, for use with manual holder, needle size 23 ga
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 24 ga, for use with manual holder
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, StableFlex, for use with manual holder
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, StableFlex, for use with autosampler
Supelco
SPME Sampling Stand, for use with 15 mL vials
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, for use with manual holder, StableFlex fiber