
Wijs (iodine monochloride) solution | Chloroiodide | 7790-99-0 solution

Synonym(s):
Iodine monochloride solution, Chloroiodide solution, Wijs solution
Linear Formula:
ICl
CAS Number:
Molecular Weight:
162.36
Products
Brand
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing