![Typhaneoside phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/382/256/6b03b583-3fec-4df5-8474-038961569dd9/640/6b03b583-3fec-4df5-8474-038961569dd9.png)
Typhaneoside
![Typhaneoside phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/382/256/6b03b583-3fec-4df5-8474-038961569dd9/640/6b03b583-3fec-4df5-8474-038961569dd9.png)
Synonym(s):
3-[(O-6-Deoxy-α-L-mannopyranosyl-(1→2)-O-[6-deoxy-a-L-mannopyranosyl-(1→6)]-β-D-glucopyranosyl)oxy]-5,7-dihydroxy-2-(4-hydroxy-3-methoxyphenyl)-4H-1-benzopyran-4-one, Isorhamnetin 3-(2′′-rhamnosylrutinoside)
Empirical Formula (Hill Notation):
C34H42O20
CAS Number:
Molecular Weight:
770.69
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing