Skip to Content
Merck
TSKgel® SP-SW Guardgel Kit phase sulfonic acid, L × I.D. 2.5 cm × 6 mm, 5 μm particle size

TSKgel® SP-SW (5 µm) HPLC Columns

TSKgel® SP-SW Guardgel Kit phase sulfonic acid, L × I.D. 2.5 cm × 6 mm, 5 μm particle size
Synonym(s):
TSKgel® Cation Exchange HPLC Guard Kit

Products

Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1