![Triglycine Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/186/276/42ce21b3-2662-42d5-8ae3-abdbf6630325/640/42ce21b3-2662-42d5-8ae3-abdbf6630325.png)
Triglycine
![Triglycine Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/186/276/42ce21b3-2662-42d5-8ae3-abdbf6630325/640/42ce21b3-2662-42d5-8ae3-abdbf6630325.png)
Synonym(s):
Gly-Gly-Gly, Glycyl-glycyl-glycine, Triglycine
Linear Formula:
NH2CH2CONHCH2CONHCH2COOH
CAS Number:
Molecular Weight:
189.17
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing