Skip to Content
Merck
Trifluoperazine hydrochloride European Pharmacopoeia (EP) Reference Standard

Trifluoperazine hydrochloride

Trifluoperazine hydrochloride European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Trifluoperazine dihydrochloride, 10-[3-(4-Methylpiperazin-1-yl)propyl]-2-(trifluoromethyl)-10H-phenothiazine dihydrochloride
Empirical Formula (Hill Notation):
C21H24F3N3S · 2HCl
CAS Number:
Molecular Weight:
480.42

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade