Torsemide Related Compound C
Synonym(s):
N-[(Ethylamino)carbonyl]-4-[(3-methylphenyl)amino]-3-pyridinesulfonamide
Empirical Formula (Hill Notation):
C15H18N4O3S
CAS Number:
Molecular Weight:
334.39
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2