Tolmetin sodium
Synonym(s):
Tolmetin sodium salt dihydrate, 1-Methyl-5-(p-toluoyl)pyrrole-2-acetic acid sodium salt dihydrate
Linear Formula:
C15H14NO3Na · 2H2O
CAS Number:
Molecular Weight:
315.30
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing