Skip to Content
Merck
vibrant-m

Thioridazine

vibrant-m
Synonym(s):
10-[2-(1-Methyl-2-piperidinyl)ethyl]-2-(methylthio)-10H-phenothiazine
Empirical Formula (Hill Notation):
C21H26N2S2
CAS Number:
Molecular Weight:
370.57

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing