![Sulindac for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/437/386/5fbd2ab1-f693-4e4b-9c47-fa41c2892a88/640/5fbd2ab1-f693-4e4b-9c47-fa41c2892a88.png)
Sulindac for system suitability
![Sulindac for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/437/386/5fbd2ab1-f693-4e4b-9c47-fa41c2892a88/640/5fbd2ab1-f693-4e4b-9c47-fa41c2892a88.png)
Synonym(s):
Sulindac, (Z)-5-Fluoro-2-methyl-1-[p-(methylsulfinyl)benzylidene]indene-3-acetic acid
Empirical Formula (Hill Notation):
C20H17FO3S
CAS Number:
Molecular Weight:
356.41
Products
Formula Weight
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing