![Sucrose monodecanoate](/deepweb/assets/sigmaaldrich/product/structures/155/179/044b77f2-ec3c-4d28-a4e3-a65c4ea4ccfb/640/044b77f2-ec3c-4d28-a4e3-a65c4ea4ccfb.png)
Sucrose monodecanoate
![Sucrose monodecanoate](/deepweb/assets/sigmaaldrich/product/structures/155/179/044b77f2-ec3c-4d28-a4e3-a65c4ea4ccfb/640/044b77f2-ec3c-4d28-a4e3-a65c4ea4ccfb.png)
Synonym(s):
Capric acid sucrose ester, Sucrose monocaprate, n-Decanoylsucrose
Empirical Formula (Hill Notation):
C22H40O12
Products
Product Category
Brand
Formula Weight
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing