![Strophanthidin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/317/012/711fce06-f37c-42ea-b4f8-dff7a0ef25cf/640/711fce06-f37c-42ea-b4f8-dff7a0ef25cf.png)
Strophanthidin
![Strophanthidin phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/317/012/711fce06-f37c-42ea-b4f8-dff7a0ef25cf/640/711fce06-f37c-42ea-b4f8-dff7a0ef25cf.png)
Synonym(s):
3β,5,14-Trihydroxy-19-oxo-5β,20(22)-cardenolide, Apocymarin, Convallatoxigenin, Corchorin, Corchoside A aglycone, Corchsularin, Cymarigenen, Erysimupicrone, K-Strophanthidin
Empirical Formula (Hill Notation):
C23H32O6
CAS Number:
Molecular Weight:
404.50
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing