Skip to Content
Merck
Spermine analytical standard

Spermine

Spermine analytical standard
Synonym(s):
N,N′-Bis(3-aminopropyl)-1,4-diaminobutane, Gerontine, Musculamine, Neuridine
Linear Formula:
NH2(CH2)3NH(CH2)4NH(CH2)3NH2
CAS Number:
Molecular Weight:
202.34
EC Number:

Products

Brand
Biological Source
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing