Skip to Content
Merck
S(−)-Nicotine solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

S(−)-Nicotine solution

S(−)-Nicotine solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Synonym(s):
(−)-Nicotine solution
Empirical Formula (Hill Notation):
C10H14N2
CAS Number:
Molecular Weight:
162.23

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Application
Package Size
Reference Material Type
Special Grade
Technique