Salidroside
Synonym(s):
2-(4-Hydroxyphenyl)ethyl β−D-glucopyranoside, p-Hydroxyphenethyl glucopyranoside, Rhodioloside, Rhodosin, Tyrosol a-(β-D-glucopyranoside)
Empirical Formula (Hill Notation):
C14H20O7
CAS Number:
Molecular Weight:
300.30
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Agency
Format
Research Area
Quality Segment
Application
Optical Isomer
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing