Riluzole Related Compound A
Synonym(s):
4-(Trifluoromethoxy)aniline, α,α,α-Trifluoro-p-anisidine
Linear Formula:
CF3OC6H4NH2
CAS Number:
Molecular Weight:
177.12
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2