Skip to Content
Merck
Rhodamine B (C.I.45170) for microscopy

Rhodamine B (C.I.45170)

Rhodamine B (C.I.45170) for microscopy
Synonym(s):
Rhodamine B (C.I.45170)
Empirical Formula (Hill Notation):
C28H31ClN2O3
CAS Number:
Molecular Weight:
479.01

Products

Brand
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing