(R)-4-Boc-morpholine-3-carboxylic acid
Empirical Formula (Hill Notation):
C10H17NO5
CAS Number:
Molecular Weight:
231.25
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing