Skip to Content
Merck
(R)-(−)-3-Bromo-2-methyl-1-propanol 97%

(R)-(−)-3-Bromo-2-methyl-1-propanol

(R)-(−)-3-Bromo-2-methyl-1-propanol 97%
Linear Formula:
BrCH2CH(CH3)CH2OH
CAS Number:
Molecular Weight:
153.02

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing