Skip to Content
Merck
p-Xylene-(dimethyl-d6) ≥98 atom % D, ≥99% (CP)

p-Xylene-(dimethyl-d6)

p-Xylene-(dimethyl-d6) ≥98 atom % D, ≥99% (CP)
Synonym(s):
1,4-Dimethyl-d6-benzene
Linear Formula:
C6H4(CD3)2
CAS Number:
Molecular Weight:
112.20

Products

Product Category
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Isotope
Showing 1-1 of 1
Compare
Product Number
Description
Pricing