![Pseudolaric Acid B phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/426/394/75f62852-6cdd-46ad-b679-0bba3d9dbf88/640/75f62852-6cdd-46ad-b679-0bba3d9dbf88.png)
Pseudolaric Acid B
![Pseudolaric Acid B phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/426/394/75f62852-6cdd-46ad-b679-0bba3d9dbf88/640/75f62852-6cdd-46ad-b679-0bba3d9dbf88.png)
Synonym(s):
(-)-Pseudolaric acid B, O-Acetylpseudolaric acid C, Pseudolarix acid B
Empirical Formula (Hill Notation):
C23H28O8
CAS Number:
Molecular Weight:
432.46
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing