Skip to Content
Merck
Pravastatin Sodium certified reference material, pharmaceutical secondary standard

Pravastatin Sodium

Pravastatin Sodium certified reference material, pharmaceutical secondary standard
Synonym(s):
Pravastatin sodium salt, (βRR,1S,2S,6S,8S,8aR)-1,2,6,7,8,8a-Hexahydro-β,,6-trihydroxy-2-methyl-8-[(2S)-2-methyl-1-oxobutoxy]-1-naphthaleneheptanoic acid monosodium salt
Empirical Formula (Hill Notation):
C23H35NaO7
CAS Number:
Molecular Weight:
446.51

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade